Learn More
Gilac™ HACCP Cook Chill Crate
Description
- 600 x 400 perforated crate.
- Ideal for commercial kitchen use and the cold chain process.
- Provides ventilation.
- Stacking and nesting when half turned.
- Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
- Crates available in 5 HACCP colors.
- Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
- Use with a water erasable marking pen.
Specifications
Specifications
| Capacity (English) | 7.1 gal. |
| Capacity (Metric) | 27 L |
| Closure Type | None |
| Color | Yellow |
| Material | HDPE |
| Shape | Rectangular |
| Product Type | Crate |
| Dimensions (L x W x H) | 600 x 400 x 146 mm |
| Length (Metric) | 600 mm |
| Length (English) | 23.6 in. |
| Show More |