missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H1FOO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116253
This item is not returnable.
View return policy
Description
H1FOO Polyclonal Antibody for Western Blot
Eukaryotic Histones are basic and water soluble nuclear proteins that form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fibers. Two molecules of each of the four core Histones (Histone H2A, H2B, H3, and H4) form the octamer, which consists of two H2A-H2B dimers and two H3-H4 dimers that are nearly symmetrical by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene, that interacts with linker DNA between nucleosomes and mediates compaction into higher order chromatin. H1FOO (H1 histone family, member O, oocyte-specific), also known as OSH1, is a 346 amino acid oocyte-specific Histone that localizes to both the nucleus and the cytoplasm. Expressed as multiple alternatively spliced isoforms, H1FOO is thought to play an important role in gene control during oogenesis and early embryogenesis and is crucial for meiotic maturation of germinal vesicle-stage oocytes.
Specifications
| H1FOO | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IZA3 | |
| H1FOO | |
| KLH conjugated synthetic peptide between 138-167 amino acids from the Central region of human H1FOO. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| H1FOO | |
| C86609; H1 histone family member O, oocyte specific; H1 histone family, member O, oocyte-specific; H1.8; H1fo; H1foo; H1OO; Histone H1oo; oocyte-specific histone H1; Oocyte-specific linker histone H1; OSH1; RGD1566236 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 132243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction