missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H-cadherin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536237
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 78, 105 and 130 kDa. Purity is >95% by SDS-PAGE.
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| H-cadherin | |
| Polyclonal | |
| Unconjugated | |
| CDH13 | |
| 4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1012, 12554, 192248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55290, Q9WTR5 | |
| CDH13 | |
| Synthetic peptide corresponding to amino acids 330-380 of human CDH13. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction