missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Kk Mouse anti-Mouse, BV605, Clone: 36-7-5, BD Biosciences™
Description
H-2Kk Monoclonal antibody specifically detects H-2Kk in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | H-2Kk |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 36-7-5 |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | H-2Kk; H2-Kk; Histocompatibility-2Kk |
| Host Species | Mouse |
| Immunogen | A.AL mouse splenocytes |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?