Learn More
Tocris Bioscience™ GYY 4137 morpholine salt
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GYY 4137 morpholine salt is a slow-releasing H2S donor. Exhibits vasodilator and antihypertensive activity. Activity causes slow dilation of blood vessels in vitro and in vivo; does not influence vascular smooth muscle cell viability in culture. Water-soluble.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | GYY 4137 morpholine salt |
| Molecular Weight (g/mol) | 376.47 |
| Inhibitors | GYY 4137 morpholine salt |
For Research Use Only
Product Content Correction
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido