Learn More
Tocris Bioscience™ GYY 4137 morpholine salt
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Beskrivelse
GYY 4137 morpholine salt is a slow-releasing H2S donor. Exhibits vasodilator and antihypertensive activity. Activity causes slow dilation of blood vessels in vitro and in vivo; does not influence vascular smooth muscle cell viability in culture. Water-soluble.
Tekniske data
Tekniske data
| Quantity | 10 mg |
| Product Type | GYY 4137 morpholine salt |
| Molecular Weight (g/mol) | 376.47 |
| Inhibitors | GYY 4137 morpholine salt |
For Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion