missing translation for 'onlineSavingsMsg'
Learn More

GW806742X, MedChemExpress

Product Code. 16485456
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485456

Brand: Medchem Express HY112292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW806742X, an ATP mimetic and a potent MLKL (Mixed Lineage Kinase Domain-Like protein) inhibitor, binds the MLKL pseudokinase domain with a Kd of 9.3 μM. GW806742X has activity against VEGFR2 (IC50=2 nM). GW806742X retards MLKL membrane translocation and inhibits necroptosis.

GW806742X, an ATP mimetic and a potent MLKL (Mixed Lineage Kinase Domain-Like protein) inhibitor, binds the MLKL pseudokinase domain with a Kd of 9.3 μM. GW806742X has activity against VEGFR2 (IC50=2 nM). GW806742X retards MLKL membrane translocation and inhibits necroptosis.

Specifications

Chemical Name or Material GW806742X
CAS 579515-63-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H22F3N7O4S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (174.35 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC(NC2=NC=CC(N(C)C3=CC=C(NC(NC4=CC=C(OC(F)(F)F)C=C4)=O)C=C3)=N2)=C1)(N)=O
Molecular Weight (g/mol) 573.55
Formula Weight 573.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.