missing translation for 'onlineSavingsMsg'
Learn More

GW4869, MedChemExpress

Product Code. p-200098453
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487748

Brand: Medchem Express HY19363

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release.

GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release.

Specifications

Chemical Name or Material GW4869
Physical Form Solid
CAS 6823-69-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H30Cl2N6O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 0.1 mg/mL (0.17 mM; Need ultrasonic) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;adjust pH to 2 with HCl) (insoluble) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(NC1=CC=C(C2=NCCN2)C=C1)/C=C/C3=CC=C(/C=C/C(NC4=CC=C(C5=NCCN5)C=C4)=O)C=C3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 577.5
Formula Weight 577.5
Percent Purity 95.57%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.