missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Carbonate, 99+%

Product Code. p-139602 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10214870

Brand: Thermo Scientific Acros 120222500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-85-1 | 0·5 H2CO3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-85-1
Molecular Formula 0·5 H2CO3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00013029

Specifications

Melting Point 267.0°C to 270.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula [C(NH2)3]2CO3
Quantity 250 g
Beilstein 03,82
Solubility Information Solubility in water: 450g/L (20 c). Other solubilities: slightly soluble in alcohol and acetone
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine carbonate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Guanidine carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.