missing translation for 'onlineSavingsMsg'
Learn More

Guaiol, MedChemExpress

Product Code. 16421738
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16421738

Brand: Medchem Express HYN3980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guaiol is a sesquiterpene alcohol that has been found in several traditional Chinese medicinal plants and has antiproliferative, pro-autophagic, insect repellent, and insecticidal biological activities.

Guaiol is a sesquiterpene alcohol that has been found in several traditional Chinese medicinal plants and has antiproliferative, pro-autophagic, insect repellent, and insecticidal biological activities.

Specifications

Chemical Name or Material Guaiol
Physical Form Solid
CAS 489-86-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H26O
For Use With (Application) COVID-19-immunoregulation
Synonym Champacol Guaiac alcohol
Solubility Information DMSO : 33.33 mg/mL (149.89 mM; Need ultrasonic)
SMILES C[C@@H]1C2=C([C@H](CC[C@@H](C(C)(O)C)C2)C)CC1
Molecular Weight (g/mol) 222.37
Formula Weight 222.37
Percent Purity 98.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.