missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C4 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
Description
GTF3C4 Monoclonal antibody specifically detects GTF3C4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | GTF3C4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4A3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | GTF3C4 |
| Gene Accession No. | Q9UKN8 |
| Gene Alias | 5330400C03; AI426938; AU014771; AU017413; general transcription factor 3C 4; general transcription factor 3C polypeptide 4; general transcription factor IIIC 4; general transcription factor IIIC subunit 4; general transcription factor IIIC, polypeptide 4; general transcription factor IIIC, polypeptide 4, 90kDa; Gtf3c4; KAT12; TF3C-delta; TFIII90; TFIIIC 90 kDa subunit; TFIIIC290; TFIIIC90; TFIIICDELTA; transcription factor IIIC 90 kDa subunit; Transcription factor IIIC subunit delta |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?