missing translation for 'onlineSavingsMsg'
Learn More

GSK9311, MedChemExpress

Product Code. p-200098311
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494426

Brand: Medchem Express HY100729

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK9311, a less active analogue of GSK6853, can be used as a negative control. GSK9311 inhibits BRPF bromodomain with pIC50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively.

GSK9311, a less active analogue of GSK6853, can be used as a negative control. GSK9311 inhibits BRPF bromodomain with pIC50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively.

Specifications

Chemical Name or Material GSK9311
CAS 1923851-49-3
Color White
Purity Grade Notes Research
Molecular Formula C24H31N5O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (228.56 mM; Need ultrasonic)
SMILES C[C@@H]1CNCCN1C2=C(N(CC)C(C3=CC=CC=C3OC)=O)C=C(C(N4C)=C2)N(C)C4=O
Molecular Weight (g/mol) 437.53
Formula Weight 437.53
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.