missing translation for 'onlineSavingsMsg'
Learn More

GSK8612, MedChemExpress

Product Code. 16417267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417267 5 mg 5mg
16454548 10 mg 10mg
16480847 50 mg 50mg
16493116 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16417267 Supplier Medchem Express Supplier No. HY111941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1.

GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1.

Specifications

Chemical Name or Material GSK8612
CAS 2361659-62-1
Color White
Purity Grade Notes Research
Molecular Formula C17H17BrF3N7O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (240.23 mM)
SMILES NS(C1=CC=C(CNC2=C(Br)C=NC(NC3=CN(CC(F)(F)F)N=C3C)=N2)C=C1)(=O)=O
Molecular Weight (g/mol) 520.33
Formula Weight 520.33
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.