missing translation for 'onlineSavingsMsg'
Learn More

GSK682753A, MedChemExpress

Product Code. 16449836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449836

Brand: Medchem Express HY101192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 (EBI2) with an IC50 of 53.6 nM.

GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 (EBI2) with an IC50 of 53.6 nM.

Specifications

Chemical Name or Material GSK682753A
Physical Form Powder
CAS 1334294-76-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H21Cl3N2O3
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 100 mg/mL (208.43 mM)
SMILES O=C1OC2(CCN(C(/C=C/C3=CC=C(Cl)C=C3)=O)CC2)CN1CC4=CC=C(Cl)C(Cl)=C4
Molecular Weight (g/mol) 479.78
Formula Weight 479.78
Percent Purity 99.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.