missing translation for 'onlineSavingsMsg'
Learn More

GSK2981278, MedChemExpress

Product Code. 16400368
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400368

Brand: Medchem Express HY19770

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2981278 is a potent and selective RORγ inverse agonist. GSK2981278 inhibits activation of the il17 promoter and interferes RORγ-DNA binding.

GSK2981278 is a potent and selective RORγ inverse agonist. GSK2981278 inhibits activation of the il17 promoter and interferes RORγ-DNA binding.

Specifications

Chemical Name or Material GSK2981278
CAS 1474110-21-8
Color White
Purity Grade Notes Research
Molecular Formula C25H35NO5S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (216.63 mM)
SMILES CC(C)CN(S(=O)(C1=CC=C(OCC2CCOCC2)C(CO)=C1)=O)C3=CC=C(CC)C=C3
Molecular Weight (g/mol) 461.61
Formula Weight 461.61
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.