missing translation for 'onlineSavingsMsg'
Learn More

GSK2837808A, MedChemExpress

Product Code. p-200098456
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463118

Brand: Medchem Express HY100681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2837808A is a potent and selective lactate dehydrogenase A (LDHA) inhibitor with IC50s of 2.6 and 43 nM for hLDHA and hLDHB, respectively.

GSK2837808A is a potent and selective lactate dehydrogenase A (LDHA) inhibitor with IC50s of 2.6 and 43 nM for hLDHA and hLDHB, respectively.

Specifications

Chemical Name or Material GSK2837808A
CAS 1445879-21-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H25F2N5O7S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (76.97 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(OC2=CC(F)=CC(F)=C2)=CC(NC3=C(S(=O)(NC4CC4)=O)C=NC5=CC(C6=CN=C(OC)N=C6OC)=CC=C35)=C1
Molecular Weight (g/mol) 649.62
Formula Weight 649.62
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.