Learn More
GSK2656157, MedChemExpress
Product Code. 16472027
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472027 50 mg 50mg
16465878 10 mg 10mg
16406346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16472027 Supplier Medchem Express Supplier No. HY13820
MedChemExpress GSK2656157 is a selective and ATP-competitive inhibitor of protein kinase R (PKR)-like endoplasmic reticulum kinase (PERK) with an IC50 of 0.9 nM.
GSK2656157 is a selective and ATP-competitive inhibitor of protein kinase R (PKR)-like endoplasmic reticulum kinase (PERK) with an IC50 of 0.9 nM.
Specifications
Chemical Name or Material GSK2656157
CAS 1337532-29-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H21FN6O
Quantity 50 mg
Solubility Information 1M HCl : 100 mg/mL (240.12 mM; ultrasonic and adjust pH to 1 with HCl) ∣DMSO : 8.33 mg/mL (20.00 mM; Need ultrasonic)
SMILES CC1=CC=CC(CC(N2CCC3=C2C=CC(C4=CN(C)C5=NC=NC(N)=C54)=C3F)=O)=N1
Molecular Weight (g/mol) 416.45
Formula Weight 416.45
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease