missing translation for 'onlineSavingsMsg'
Learn More

GSK2636771, MedChemExpress

Product Code. 16498977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498977

Brand: Medchem Express HY15245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2636771 is a potent, selective and orally bioavailable inhibitor of PI3Kβ with a Ki of 0.89 nM and an IC50 of 5.2 nM, showing 900-fold selectivity over p110α and p110γ, and 10-fold selectivity over p110δ isoforms.

GSK2636771 is a potent, selective and orally bioavailable inhibitor of PI3Kβ with a Ki of 0.89 nM and an IC50 of 5.2 nM, showing 900-fold selectivity over p110α and p110γ, and 10-fold selectivity over p110δ isoforms.

Specifications

Chemical Name or Material GSK2636771
CAS 1372540-25-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H22F3N3O3
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (57.68 mM; Need ultrasonic)
SMILES CC1=NC2=C(C(O)=O)C=C(C=C2N1CC3=CC=CC(C(F)(F)F)=C3C)N4CCOCC4
Molecular Weight (g/mol) 433.42
Formula Weight 433.42
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.