missing translation for 'onlineSavingsMsg'
Learn More

GSK1940029, MedChemExpress

Product Code. p-200101924
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493837

Brand: Medchem Express HY19762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK1940029 (SCD inhibitor 1) is a stearoyl-coa desaturase (SCD) inhibitor extracted from patent WO/2009060053 A1, compound example 16.

GSK1940029 (SCD inhibitor 1) is a stearoyl-coa desaturase (SCD) inhibitor extracted from patent WO/2009060053 A1, compound example 16.

Specifications

Chemical Name or Material GSK1940029
CAS 1150701-66-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H16Cl2N4O2
Quantity 50 mg
Synonym SCD inhibitor 1
Solubility Information DMSO : ≥ 32 mg/mL (81.79 mM)
SMILES ClC(C=CC(CN1N=NC(C(NC2=CC=C(CO)C=C2)=O)=C1C)=C3)=C3Cl
Molecular Weight (g/mol) 391.25
Formula Weight 391.25
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.