missing translation for 'onlineSavingsMsg'
Learn More

GSK 3 Inhibitor IX, MedChemExpress

Product Code. 16464058
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464058

Brand: Medchem Express HY10580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK 3 Inhibitor IX (6-Bromoindirubin-3'-oxime; BIO) is a potent, selective, reversible and ATP-competitive inhibitor of GSK-3α/β and CDK1-cyclinB complex with IC50s of 5 nM/320 nM/80 nM for (GSK-3α/β)/CDK1/CDK5, respectively.

GSK 3 Inhibitor IX (6-Bromoindirubin-3'-oxime

Specifications

Chemical Name or Material GSK 3 Inhibitor IX
CAS 667463-62-9
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C16H10BrN3O2
Quantity 10 mg
Synonym 6-Bromoindirubin-3'-oxime BIO MLS 2052
Solubility Information DMSO : ≥ 23 mg/mL (64.58 mM)
SMILES O/N=C(C1=CC=CC=C1N2)/C2=C3C(NC4=C/3C=CC(Br)=C4)=O
Molecular Weight (g/mol) 356.17
Formula Weight 356.17
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.