Learn More
Tocris Bioscience™ GSK 2837808A
Potent and selective LDHA inhibitor
Brand: Tocris Bioscience™ 5189/10
This item is not returnable.
View return policy
Description
GSK 2837808A is a potent and selective lactate dehydrogenase A and B (LDHA and LDHB) inhibitor (IC50 values are 2.6 and 43 nM for LDHA and LDHB respectively). Inhibits lactate production in selected cancer cell lines. Reduces glucose uptake and enhances mitochondrial oxygen consumption in Snu398 hepatocellular carcinoma cells. Inhibits proliferation and induces apoptosis in Snu398 cells. Inhibits transcription of histone 2B (H2B) gene in HCT116 and NCM460 cells. Cell permeable.Specifications
| 98% | |
| 1445879-21-9 | |
| 10 mg |
| 3-[[3-[(Cyclopropylamino)sulfonyl]-7-(2,4-dimethoxy-5-pyrimidinyl)-4-quinolinyl]amino]-5-(3,5-difluorophenoxy)benzoic acid | |
| -20°C |