missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
| Antigen | GS28 |
| Applications | Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GOSR1 |
| Gene Accession No. | O88630, O95249, Q62931 |
| Gene Alias | 28 kDa cis Golgi SNARE p28, 28 kDa Golgi SNARE protein, GOLIM2, GOS 28, GOS28, GOS28/P28, GOSR1, GS28, P28 |
| Gene Symbols | Gosr1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?