Learn More
Invitrogen™ GRK2 Monoclonal Antibody (5D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA12013
Description
MA1-2013 detects the beta andrenergic receptor kinase 1 in human, mouse and rat samples. MA1-2013 has been successfully used in Western blot, immunocytochemistry, immunofluoresence, immunoprecipitation, and ELISA procedures. By Western blot this antibody detects a ~30 kDa protein representing the beta andrenergeic receptor kinase 1 in human HeLa cells. In immunocytochemistry procedures MA1-2013 recognizes the beta andrenergic receptor kinase 1 in HeLa, Hek-293 and 3T3 cells. Immunofluoresence in HeLa cells yeilds predominantely cytoplasmic membrane associated staining with some staining in the ruffles. The MA1-2013 immunogen is recombinant beta adrenergic receptor kinase 1.
ADRBK1 (GRK2) is a serine/threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs). G protein-coupled receptor kinases (GRKs) are important regulators of G protein-coupled receptors(GPCRs). GRK2 (83 kDa), one of six members of this family that have been identified, is ubiquitously expressed in mammals. After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs (e.g., a2-adrenergic receptor) are phosphorylated by GRKs. Internalization of the GPCRs, regulated by beta-arrestin-1, leads to activation of the Ras - Raf - ERK1&2 signaling pathway. GRK2 activity is tightly controlled by different mechanisms including phosphorylation by kinases such as PKC, Src and ERK1&2, as well as interaction with various proteins. ERK phosphorylates and thus inactivates GRK2 on serine 670 in a negative feedback mechanism.
Specifications
| GRK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P25098, P26817, Q99MK8 | |
| GRK2 | |
| Recombinant beta adrenergic receptor kinase 1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5D5 | |
| Unconjugated | |
| GRK2 | |
| ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; EC 2.7.1.126; G protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2; Grk2; GRK-2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 110355, 156, 25238 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.