Learn More
Griffin Education™ Mass Set
Click to view available options
:
Mass: 100g; Packaging Quantity: 4/PK
Specifications
Specifications
| Product Type | Mass Set |
| Mass | 100 g |
| Includes | 10g Steel wire mass hanger cast alloy, hook and nine Alloy 10g masses |
| Quantity | 4/Pk. |
| Packaging Quantity | 4/Pk. |