missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRG (Groucho homolog) Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76645MFV500
This item is not returnable.
View return policy
Description
GRG (Groucho homolog) Polyclonal antibody specifically detects GRG (Groucho homolog) in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| GRG (Groucho homolog) | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human AES. The immunogen is located within the last 50 amino acids of AES. Amino Acid Squence: GHDGDTHQEDDGEKSD | |
| 0.1 mL | |
| Apoptosis, Signal Transduction, Wnt Signaling Pathway | |
| 166 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction