Learn More
Deutsch Neumann™ Grey Natural Rubber Spacer
Click to view available options
Height (Metric):
20 mm
24 mm
Quantity:
10/Pk.
Unit Size:
Pack of 10
Specifications
Specifications
| Color | Gray |
| Material | Natural Rubber |
| No. per Pack | 10Pack |
| Quantity | 10/Pk. |
| Height (Metric) | 20 mm |