missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grazoprevir potassium salt, MedChemExpress
Description
Grazoprevir potassium salt (MK-5172 potassium salt) is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across genotypes and resistant variants, with Kis of 0.01 nM (gt1b), 0.01 nM (gt1a), 0.08 nM (gt2a), 0.15 nM (gt2b), 0.90 nM (gt3a), respectively. Grazoprevir potassium salt inhibits SARS-CoV-2 3CLpro activity.
Specifications
Specifications
| Chemical Name or Material | Grazoprevir potassium salt |
| CAS | 1206524-86-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H49KN6O9S |
| Quantity | 50 mg |
| Synonym | MK-5172 potassium salt |
| Solubility Information | DMSO : ≥ 100 mg/mL (124.23 mM) |
| SMILES | COC1=CC2=C(N=C(CCCCC[C@@H]3C[C@H]3OC4=O)C(O[C@H]5CN(C([C@H](C(C)(C)C)N4)=O)[C@H](C(N[C@@]([C@@H]6C=C)(C6)C([N-]S(C7CC7)(=O)=O)=O)=O)C5)=N2)C=C1.[K+] |
| Molecular Weight (g/mol) | 804.99 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?