missing translation for 'onlineSavingsMsg'
Learn More

Grazoprevir hydrate, MedChemExpress

Product Code. 16472537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472537 50 mg 50mg
16429027 5 mg 5mg
16426438 10 mg 10mg
16416726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472537 Supplier Medchem Express Supplier No. HY15298B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Grazoprevir hydrate (MK-5172 hydrate) is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across genotypes and resistant variants, with Kis of 0.01 nM (gt1b), 0.01 nM (gt1a), 0.08 nM (gt2a), 0.15 nM (gt2b), 0.90 nM (gt3a), respectively. Grazoprevir hydrate inhibits SARS-CoV-2 3CLpro activity.

Grazoprevir hydrate (MK-5172 hydrate) is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across genotypes and resistant variants, with Kis of 0.01 nM (gt1b), 0.01 nM (gt1a), 0.08 nM (gt2a), 0.15 nM (gt2b), 0.90 nM (gt3a), respectively. Grazoprevir hydrate inhibits SARS-CoV-2 3CLpro activity.

Specifications

Chemical Name or Material Grazoprevir hydrate
CAS 1350462-55-3
Color White
Purity Grade Notes Research
Molecular Formula C38H52N6O10S
Quantity 50 mg
Synonym MK-5172 hydrate
Solubility Information DMSO : 50 mg/mL (63.70 mM; Need ultrasonic)
SMILES COC1=CC2=C(N=C(CCCCC[C@@H]3C[C@H]3OC4=O)C(O[C@H]5CN(C([C@H](C(C)(C)C)N4)=O)[C@H](C(N[C@@]([C@@H]6C=C)(C6)C(NS(C7CC7)(=O)=O)=O)=O)C5)=N2)C=C1.O
Molecular Weight (g/mol) 784.92
Formula Weight 784.92
Percent Purity 99.1%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.