Learn More
Grant Instruments™ T100IN Inspissator Control Unit
Product Code. 15231176
Click to view available options
For Use With (Equipment):
For Inspissator
Unit Size:
Each
Product Code. 15231176
Brand: Grant Instruments™ T100IN
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Temperature Range: Ambient +5°C to 100°C
  • Clear 4 digit display - easy to read from a distance for instant re-assurance.
  • Digital temperature control
  • Standard temperature: 85°C
  • temperature range ambient +5°C to 100°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For Inspissator
Temperature Range (Metric) Ambient +5° to 100°C
Type Control Unit