Learn More
KARTELL Graduated Burette
Click to view available options
Capacity (Metric):
10 mL
50 mL
Unit Size:
Each
Description
- High precison plastic burettes (according to DIN 12.700 standards - Class B), particularly useful for routine work to eliminate the danger of glass breakages.
- The combination of different plastic materials (PMMA, PTFE, FEP) provides good resistance to chemicals.
- The stopcock assembly is moulded in one piece, with FEP key; requiring no lubricant and providing a leak-proof seal.
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Length (Metric) | 740 mm |
| Material | Acrylic |
| Graduations | 0.1 mL |
| Length (English) | 29.13 in. |
| Stopcock Material | PTFE |
| Type | Graduated Buret |