missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 3 Qualitative High Purity Lab Filter Papers

For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis.

Brand:  Munktell Filtrak™ 110063

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 11313074

  • 10.25 EUR / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Made of pure cotton linters
  • For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
  • Basis Weight: 90g/m2
Specifications

Specifications

Disc
<10 μm
3
Cotton Linters
11s/10 mL
55 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers