missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade 1F Qualitative High Purity Lab Filter Papers
For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate.
Brand: Munktell Filtrak™ 110025
Additional Details : Weight : 0.68400kg
Description
- Made of pure cotton linters
- For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate
- Basis Weight: 80g/m2
Specifications
Disc | |
5 to 6 μm | |
1F |
Cotton Linters | |
29s/10 mL | |
32 cm |