missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR98 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599894
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR98.
This gene encodes a member of the G-protein coupled receptor superfamily. The encoded protein contains a 7-transmembrane receptor domain, binds calcium and is expressed in the central nervous system. Mutations in this gene are associated with Usher syndrome 2 and familial febrile seizures. Several alternatively spliced transcripts have been described.
Specifications
| GPR98 | |
| Polyclonal | |
| Unconjugated | |
| ADGRV1 | |
| 4-Feb; ADGRV1; ADGRV1 subunit alpha; ADGRV1 subunit beta; adhesion G protein-coupled receptor V1; Adhesion G-protein coupled receptor V1; FEB4; Frings; G protein-coupled receptor 98; Gpr98; G-protein coupled receptor 98; KIAA0686; KIAA1943; MASS1; Mgr1; monogenic; monogenic audiogenic seizure susceptibility protein 1; monogenic audiogenic seizure susceptibility protein 1 homolog; monogenic, audiogenic seizure susceptibility 1; monogenic, audiogenic seizure susceptibility 1 homolog; neurepin; USH2B; USH2C; Usher syndrome type-2C protein; very large G protein-coupled receptor 1; very large G-protein coupled receptor 1; VLGR1; VLGR1b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110789, 84059 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXG9 | |
| ADGRV1 | |
| A synthesized peptide derived from human GPR98(Accession Q8WXG9), corresponding to amino acid residues L2004-S2054. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction