missing translation for 'onlineSavingsMsg'
Learn More

GPR44 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25264-1-AP-20UL

 View more versions of this product

Product Code. 16823845

  • 145.00 EUR / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CRTH2 (chemoattractant receptor-homologous molecule), which is also known as the putative G protein-coupled receptor (GPR44), is expressed on Th2 cells and is a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells. CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions.
Specifications

Specifications

GPR44
Polyclonal
Unconjugated
PTGDR2
CD294, CRTH2, DL1R, DP2, G protein coupled receptor 44, GPR44, Prostaglandin D2 Receptor 2, PTGDR2
Rabbit
Antigen Affinity Chromatography
RUO
11251
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y5Y4
PTGDR2
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.