missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR43 Polyclonal Antibody

Product Code. 11536182
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11536182 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11536182 Supplier Invitrogen™ Supplier No. PA120419

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-20419 detects G Protein-Coupled Receptor GPR43 in human samples.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR43
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene FFAR2
Gene Accession No. O15552
Gene Alias fatty acid receptor 2; FFA2; FFA2R; FFAR2; free fatty acid activated receptor 2; free fatty acid receptor 2; G protein-coupled receptor 43; GPCR43; Gpr43; G-protein coupled receptor 43; leukocyte-specific STAT-induced GPCR; Lssig
Gene Symbols FFAR2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd extracellular domain of human GPR43.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2867
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.