missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR177/WLS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 550.00 EUR
Specifications
| Antigen | GPR177/WLS |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230888
|
Novus Biologicals
NBP3-35821-20ul |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227282
|
Novus Biologicals
NBP3-35821-100ul |
100 μL |
550.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR177/WLS Polyclonal antibody specifically detects GPR177/WLS in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| GPR177/WLS | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 79971 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C1orf139, DKFZp686I0788, EVIchromosome 1 open reading frame 139, FLJ23091, G protein-coupled receptor 177, GPR177wls, Integral membrane protein GPR177, MGC131760, MGC14878, MRP, Protein evenness interrupted homolog, protein wntless homolog, Putative NF-kappa-B-activating protein 373, putative NFkB activating protein 373, wntless homolog (Drosophila) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 43-232 of human GPR177/WLS (NP_001002292.3).,, Sequence:, YMSVKCVDARKNHHKTKWFVPWGPNHCDKIRDIEEAIPREIEANDIVFSVHIPLPHMEMSPWFQFMLFILQLDIAFKLNNQIRENAEVSMDVSLAYRDDAFAEWTEMAHERVPRKLKCTFTSPKTPEHEGRYYECDVLPFMEIGSVAHKFYLLNIRLPVNEKKKINVGIGEIKDIRLVGIHQNGGFTKVW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title