missing translation for 'onlineSavingsMsg'
Learn More

GPR120 modulator 1, MedChemExpress

Product Code. p-200099897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470528

Brand: Medchem Express HY50162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GPR120 modulator 1 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F1. GPR120 modulator 1 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.

GPR120 modulator 1 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F1. GPR120 modulator 1 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.

Specifications

Chemical Name or Material GPR120 modulator 1
CAS 1050506-75-6
Color White
Purity Grade Notes Research
Molecular Formula C19H16ClNO4S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (256.51 mM)
SMILES O=C(O)COC1=CC=C(OCC2=NC(C3=CC=CC=C3Cl)=CS2)C=C1C
Molecular Weight (g/mol) 389.85
Formula Weight 389.85
Percent Purity 98.56%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.