missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR120 Agonist 3, MedChemExpress
Description
GPR120 Agonist 3 is a selective Gpr120 agonist with a logEC50 of −7.62.
Specifications
Specifications
| Chemical Name or Material | GPR120 Agonist 3 |
| CAS | 1599477-75-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H23ClF3NO3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (123.20 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(O)CC(CC1)CCC21CCN(C3=CC(OC(F)(F)F)=CC=C3Cl)CC2 |
| Molecular Weight (g/mol) | 405.84 |
| Formula Weight | 405.84 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?