missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp130/IL6ST Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67766-1-IG-150UL
This item is not returnable.
View return policy
Description
The gp130/IL6ST antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human gp130/IL6ST. This antibody recognizes human antigen. The gp130/IL6ST antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| gp130/IL6ST | |
| Monoclonal | |
| 1000 μg/mL | |
| CD130; CDw130; GP130; GP130-RAPS; IL6R-beta | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 130-140 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1H7C5 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 3572 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction