Learn More
gp100 / Melanosome / PMEL17 / SILV (Melanoma Marker) Mouse, Clone: PMEL/2037, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6490-MSM7-P0
Denne vare kan ikke returneres.
Se returpolitik
Tekniske data
| gp100 / Melanosome / PMEL17 / SILV (Melanoma Marker) | |
| Cytoplasmic | |
| PMEL/2037 | |
| Unconjugated | |
| Mouse | |
| 90-100kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 6490 | |
| IgG1 κ |
| Immunohistochemistry, ELISA | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human SILV protein fragment (apprx. aa 376-502) | |
| SK-MEL-28 cells or Melanoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion