missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (SPM581), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34813F
This item is not returnable.
View return policy
Description
Golgi Complex Monoclonal specifically detects Golgi Complex in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Golgi Complex | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SU-DHL-1 large cell lymphoma cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM581 | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This monoclonal antibody recognizes Golgi complex in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the Golgi complex in human cells only. It can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells and may be used to stain Golgi complex of cells in frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. This monoclonal antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction