Learn More
3M™ GoggleGear™ 6000 Safety Goggles
Description
- For safety goggles with indirect ventilation and a 3M™ Scotchgard™ Anti-Fog and Anti-Scratch coating, use 3M™ GoggleGear™ Safety Goggles 6000 Series.
- Our large profile safety goggles feature indirect ventilation to allow air to circulate and help reduce fogging.
- The polycarbonate lens features a 3M™ Scotchgard™ Anti-Fog and Anti-Scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water*.
- Available in a range of shroud colours and two lens options.
- *3M internal testing per EN168 test method when compared with traditional anti-fog coatings.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | None |
| Product Type | Safety Goggles |
| Frame Color | Blue |
| Material | Polypropylene |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polypropylene |
| UV Protection | UV400 |
| Frame Type | Large Profile |