Learn More
Invitrogen™ Goat anti-Mouse IgM Cross-Adsorbed Secondary Antibody, DyLight™ 550
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ SA510151
Product Code. 13427307
  • 194.00 EUR / 500µg
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to human and rat.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgM Cross-Adsorbed
Polyclonal
DyLight 550
Goat
Antigen affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Mouse IgM.
500 μg
Secondary
4°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions