Learn More
KL Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of KL.
Brand: Abnova PAB6906.100ug
Description
This gene encodes a type-I membrane protein that is related to beta-glucosidases. Reduced production of this protein has been observed in patients with chronic renal failure (CRF), and this may be one of the factors underlying the degenerative processes (e.g., arteriosclerosis, osteoporosis, and skin atrophy) seen in CRF. Also, mutations within this protein have been associated with ageing and bone loss. [provided by RefSeq
Sequence: C-KRDDAKYMYYLKKSpecifications
| KL | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| KL | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9365 | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of KL. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| KL | |
| A synthetic peptide corresponding to human KL. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_004786.2, NP_710150.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only