Learn More
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, Biotin
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ 31770
Product Code. 11583330
  • 215.00 EUR / 1.50mg
Description
Product # 31770 has been successfully used in Western blot, IF, ICC, IHC and ISH applications. Product # 31770 reacts with the heavy chains of human IgG and with the light chains common to most human immunoglobulins, but does not react against non-immunoglobulin serum proteins. Based on solid-phase enzyme immunoassay, this antibody cross-reacts with horse IgG, human IgM, swine IgG (6%), cat IgG, chicken IgG, dog IgG, dog IgM, guinea pig IgG, mouse IgM, swine IgM (3%), mouse IgG, rabbit IgG, rat IgG, rat IgM (1%), and it has minimal cross-reactivity with immunoglobulins from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgG (H+L)
Polyclonal
Biotin
Goat
1.5 mg
Secondary
4°C
Liquid
ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
1.5 mg/mL
PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8
Affinity chromatography
RUO
Human
Secondary Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions