missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74988-0.5mg
113.05 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications
| IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy gamma chains on human IgG. This antibody has been pre-adsorbed against mouse IgG1 and bovine, horse, mouse, and rabbit serum proteins. | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
| Immunoassay | |
| 4.5 mg/mL | |
| O75015, O75015, O75015 | |
| Purified Human IgG, Fc fragment | |
| 0.5 mg | |
| Secondary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction