Learn More
CNN3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CNN3.
Brand: Abnova PAB18717.100ug
Description
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction. [provided by RefSeq
Sequence: C-HGEYQDDYPRDYQYSSpecifications
| CNN3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user. | |
| CNN3 | |
| CNN3 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human CNN3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CNN3. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_001830.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1266 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only