missing translation for 'onlineSavingsMsg'
Learn More

GNE-6640, MedChemExpress

Product Code. 16475556
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475556

Brand: Medchem Express HY112937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-6640 is a selective and non-covalent inhibitor of ubiquitin epecific peptidase 7 (USP7), with IC50 values of 0.75 μM, 0.43 μM, 20.3 μM and 0.23 μM for full length USP7, USP7 catalytic domain, full length USP43 and Ub-MDM2, respectively.

GNE-6640 is a selective and non-covalent inhibitor of ubiquitin epecific peptidase 7 (USP7), with IC50 values of 0.75 μM, 0.43 μM, 20.3 μM and 0.23 μM for full length USP7, USP7 catalytic domain, full length USP43 and Ub-MDM2, respectively.

Specifications

Chemical Name or Material GNE-6640
CAS 2009273-67-8
Color White
Purity Grade Notes Research
Molecular Formula C20H18N4O
Quantity 100 mg
Solubility Information DMSO : 5 mg/mL (15.13 mM; Need ultrasonic)
SMILES OC1=CC=C(C2=C(CC)C(C3=CC4=C(NN=C4)C=C3)=CN=C2N)C=C1
Molecular Weight (g/mol) 330.38
Formula Weight 330.38
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.