missing translation for 'onlineSavingsMsg'
Learn More

GNE-317, MedChemExpress

Product Code. p-200101303
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465328 10 mg 10mg
16428047 5 mg 5mg
16415976 100 mg 100mg
16401537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465328 Supplier Medchem Express Supplier No. HY12763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-317 is a PI3K/mTOR inhibitor, is able to cross the blood-brain barrier (BBB).

GNE-317 is a PI3K/mTOR inhibitor, is able to cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material GNE-317
Physical Form Solid
CAS 1394076-92-6
Color Earth Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H22N6O3S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (48.25 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=NC(N3CCOCC3)=C4C(C(C)=C(C5(OC)COC5)S4)=N2)C=N1
Molecular Weight (g/mol) 414.48
Formula Weight 414.48
Percent Purity 99.31%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.