missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAQ Antibody (GNAQ/2434), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08585AF488
This item is not returnable.
View return policy
Description
GNAQ Monoclonal antibody specifically detects GNAQ in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| GNAQ | |
| Monoclonal | |
| Alexa Fluor 488 | |
| GAQG-ALPHA-q, guanine nucleotide binding protein (G protein), q polypeptide, Guanine nucleotide-binding protein alpha-q, guanine nucleotide-binding protein G(q) subunit alpha | |
| Recombinant full-length human GNAQ protein (Uniprot: P50148) | |
| 100 μg | |
| Cardiovascular Biology | |
| 2776 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| GNAQ/2434 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction